Shaft Coupling, Stirring, Ace Glass Incorporated
Supplier: Ace Glass
8125-06 8125-08 8125-11 8125-13
89062-266EA 157.24 CAD
89062-266 89062-268 89062-270 89062-272 89062-274 89062-276 89062-278 89062-280
Shaft Coupling, Stirring, Ace Glass Incorporated
Stirrers Stirring Shafts
The coupling connects the stirring motor shaft to the reactor stirring shaft.
The coupling has a flexible neoprene rubber body with a PTFE sleeve inside the corrosion-resistant metal end. This provides angular and parallel alignment of glass or metal stirring shafts without the danger of breakage. The rubber body absorbs shock and provides quiet, vibration-free operation with great torsional stiffness. Shaft coupling measures 4" overall.