Mantle, Aluminum Housing, Solid Bottom, Ace Glass Incorporated
Supplier: Ace Glass
12053-05 12053-07 12053-13 12053-17 12053-25 12053-19 12053-21 12053-23 12053-26
89075-944EA 926.71 CAD
89075-944 89075-946 89075-948 89075-950 89075-958 89076-026 89076-028 89076-030 89084-348
Mantle, Aluminum Housing, Solid Bottom, Ace Glass Incorporated
Heaters Heating Mantles
Solid, aluminum-housed heating mantles are versatile enough to be used on lab benches, stands, or floors
They accommodate cylindrical and spherical flasks. Mantles must be operated through a temperature controller, and offer an operating temperature range of 10° to 450°C. All units are supplied with a 122 cm (4') detachable cord with interlocking connector.
Certifications: CSA approved.