Mantle, Aluminum Housing, Low Form, Solid Bottom, for Cylindrical Flasks, Ace Glass Incorporated
Supplier: Ace Glass
12053-40 12053-43 12053-45
89076-052EA 2704.86 CAD
89076-052 89076-054 89076-056
Mantle, Aluminum Housing, Low Form, Solid Bottom, for Cylindrical Flasks, Ace Glass Incorporated
Heaters Heating Mantles
Solid, aluminum housed heating mantles are designed for use with cylindrical flasks
They can be used on benches with smaller sized flasks or on floor or reactor stands with larger flasks. Mantles must be operated through a temperature controller, and offer an operating temperature range of ambient +10° to 450°C. Mantles are supplied with either a 122cm (4') detachable cord with interlocking connector, or with a terminal box for hard wiring. CSA approved.