Bearing, Trubore®, PTFE, 19 mm, Ace Glass Incorporated
Supplier: Ace Glass
8067-34 8067-38 8067-05 8067-08 8067-13 8067-30 8067-11
89061-894EA 163.24 CAD
89061-894 89061-912 89061-900 89061-910 89061-898 89061-892 89061-908
Bearing, Trubore®, PTFE, 19 mm, Ace Glass Incorporated
Stirrers Stirring Tools
Self-aligning, lubricant-free PTFE Trubore® bearing for use with a 19mm polished glass stirring shaft
Complete item consists of PTFE inner Trubore® bearing, #25 Ace-Thred glass adapter, non-flaking PTFE compression saddle with o-ring and glass-reinforced PTFE bushing and lock nut with FETFE o-ring. The lock nut permits stirring in either direction without fear of unthreading. The compression saddle with o-ring maintains constant force with little attention. Vacuum down to 1mm of Hg. Not recommended for use with precision ground glass shafts.
Ordering information: For replacement "saddle" size -210 FETFE o-rings, order 89061-012; for replacement bearing size -212 FETFE o-ring, order 89061-016. For #25 Ace-Thred adapter (only) with [ST] 45/50 joint, order 89061-902. For #25 Ace-Thred adapter (only) with [ST] 55/50 joint, order 89061-904. For #25 Ace-Thred adapter (only) with [ST] 71/60 joint, order 89061-906.