Cooler, Condensate, Rotary Evaporator, Ace Glass Incorporated
Supplier: Ace Glass
3965-36 3965-34
89214-988EA 2637.78 CAD
89214-988 89214-990
Cooler, Condensate, Rotary Evaporator, Ace Glass Incorporated
Condensers
Replacement borosilicate glass cooler for popular process-scale rotary evaporators.
Has DN40 ball joint at both ends. Available poly coated for safety, or uncoated.
Glassware set: C, RB, R
Ordering information: Includes size -217 CAPFE (silicone-encapsulated rubber) 89061-126 o-ring.