STERILE VERTEX® Polyester Pre-Wetted Wipers, Texwipe®
Supplier: Texwipe
TX3044P TX3049P TX3042P
TWTX3049PCS 347.54 CAD
TWTX3049P TWTX3044P TWTX3042P
STERILE VERTEX® Polyester Pre-Wetted Wipers, Texwipe®
Wipes Cleanroom Wipes Presaturated Cleanroom Wipes
Processed in a fully-automated, hands-free micro environment (Texwipe's unique Vertex® manufacturing system), these polyester, sealed-edge wipers are ultra clean.
TWTX3042P and TWTX3049P feature the same ultra clean wipers pre-wetted with 70% USP-grade IPA while TWTX3044P comes pre-wetted with 70% USP-grade ethyl alcohol. These are 100% continuous-filament, polyester knit fabric wipers made in the USA from virgin stock and processed on Texwipe’s Vertex® manufacturing system. The wiper’s unique design provides a very soft feel and high sorptive capacity.
Quarterly agar overlay and LAL testing assures that wipers do not contribute cytotoxic leachables and are nonpyrogenic.
The wiper's unique design provides a very soft feel and high sorptive capacity in a single ply wiper.
Ordering information: Available in either dry or pre-wetted (with 70% USP-grade isopropyl/ethyl alcohol and packaged in a standup reclosable slider bag.) Also available in Sterile Microdenier Dry Series.