Ampicillin sodium salt, powder

Supplier: Corning

61-238-RH 61-238-RM
CA45000-612EA 342.01 CAD
CA45000-612 CA45000-614
Ampicillin sodium salt, powder
Ampicillin sodium salt

Ampicillin is a Penicillin derivative that inhibits bacterial cell-wall synthesis by inactivating transpeptidases on the inner surface of the bacterial cell membrane.


A ß-lactam antibiotic with an amino group side chain attached to the penicillin structure, its mode of resistance is through the cleavage of ß-lactam ring of ampicillin by ß-lactamase.


Ampicillin is effective against Gram-positive and some Gram-negative bacteria.


Packaging: Shipping temperature: Ambient

Formula: C₁₆H₁₈N₃NaO₄S
Storage Temperature: Refrigerator
MDL Number: MFCD00064313
CAS Number: 69-52-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR