Cork Rings, Ace Glass Incorporated
Supplier: Ace Glass
11750-04 11750-06 11750-08 11750-10 11750-12 11750-02
89075-768EA 27.89 CAD
89075-768 89075-770 89075-772 89075-774 89075-776 89075-766
Cork Rings, Ace Glass Incorporated
Supports Flask Supports
Fine-composition cork rings ideally suited for general laboratory use to support flasks, beakers, dishes
Rings are 30mm (1-3/16-inch) thick. Available in a variety of sizes.