Eyesaver Drench Hose and Eyewash Combination, Model SE-920, Speakman®
Supplier: Speakman
Eyesaver®
SE-920
VF-SE-920EA 1205.89 CAD
VF-SE-920
Eyesaver Drench Hose and Eyewash Combination, Model SE-920, Speakman®
First Aid Supplies Eyewash Stations and Eyewash Bottles
Speakman® Drench hose and eyewash combinations provide great flexibility in laboratories and other industrial areas for flushing the eyes and body.
  • Hose and eyewash combination
  • Laboratory flexibility
  • For direct flushing on contaminated areas
All units come standard with a flexible hose. Model SE-920 is a wall mounted drench hose featuring one aerated spray outlet and a 6' hose. The unit also features a self-closing valve and hook for the drench hose.
Certifications: Meets ANSI 358.1.