Upchurch Scientific® Pressure Relief Valves, IDEX Health & Science
Supplier: Upchurch Scientific
P-612 U-456
21511-160EA 237.02 CAD
21511-160 21511-532
Upchurch Scientific® Pressure Relief Valves, IDEX Health & Science
Valves Non Return Valves
Prevent system over-pressurization with Upchurch Scientific® Pressure Relief Valves
To open the Pressure Relief Valve Assembly at a different pressure than 100psi (7 bar), simply combine one of the Upchurch Scientific® Replacement Back Pressure Regulator Assemblies with item 21511-160 Pressure Relief Valve Tee, or item 21528-160 for larger bore tubing and higher flow applications.
These products protect system components by diverting fluid flow automatically when inline pressure exceeds the set limit.
Certifications: ROHS certified.
Ordering information: The 21511-532 assemblyMore Product Information