N-Lauryldiethanolamine ≥95.0%

Supplier: TCI America

Synonyms: N-Dodecyldiethanolamine, N,N-Bis(2-hydroxyethyl)dodecylamine

L0020-25G L0020-5G
TCL0020-25GEA 401.32 CAD
TCL0020-25G TCL0020-5G
N-Lauryldiethanolamine ≥95.0%
N-Lauryldiethanolamine

CAS Number: 1541-67-9
MDL Number: MFCD00059002
Molecular Formula: C16H35NO2
Molecular Weight: 273.46
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 195
Specific Gravity (20/20): 0.92


Other Product Names / Synonyms: N,N-Bis(2-hydroxyethyl)dodecylamine, N-Dodecyldiethanolamine

Formula: C₁₆H₃₅NO₂
Boiling Pt: 195 °C
Density: 0.92 (20 °C)
MDL Number: MFCD00059002
CAS Number: 1541-67-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a43e19301e3f4cdb10943e88816972ee34342f9b&vid=a43e19301e3f4cdb10943e88816972ee34342f9b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR