N-Fmoc-L-alanine ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
N-(9-Fluorenylmethoxycarbonyl)-L-alanine, (-)-N-{[(9H-Fluoren-9-yl)methoxy]carbonyl}-L-alanine, Fmoc-L-alanine, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-alanine, (-)-L-Alanine, N-FMOC protected, (-)-FMOC-L-Ala-OH, (2S)-(-)-2-Aminopropanoic acid, N-FMOC protected, Fmoc-Ala-OH
Form: Crystal
Specific rotation [a]20/D: -18.5 deg (C=1, DMF)
Product Categories: Synthetic Organic Chemistry, Peptide Synthesis, Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: Fmoc-L-alanine, Fmoc-Ala-OH
Formula:
C₁₈H₁₇NO₄ MW: 311.34 g/mol |
MDL Number:
MFCD00037139 CAS Number: 35661-39-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...