cis-13-Docosenol ≥98.0%
Supplier: TCI America
D2174-1G
D2174-5G
TCD2174-001GEA
48.98
CAD
TCD2174-001G
TCD2174-5G
cis-13-Docosenol ≥98.0%
cis-13-Docosenol
CAS Number: 629-98-1
MDL Number: MFCD00055936
Molecular Formula: C22H44O
Molecular Weight: 324.59
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 225
Melting point (°C): 33
Product Categories: Bioscience, Higher Fatty Acids & Higher Alcohols
Other Product Names / Synonyms: Erucyl Alcohol
Formula:
C₂₂H₄₄O Boiling Pt: 225 °C Melting Pt: 33 °C |
MDL Number:
MFCD00055936 CAS Number: 629-98-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...