5-Chlorovaleric acid ≥96.0% (by GC, titration analysis)

Supplier: TCI America
Danger

C0309-25G C0309-5G
TCC0309-025GEA 71.1 CAD
TCC0309-025G TCC0309-005G
5-Chlorovaleric acid ≥96.0% (by GC, titration analysis)
5-Chlorovaleric acid

CAS Number: 1119-46-6
MDL Number: MFCD00004415
Molecular Formula: C5H9ClO2
Molecular Weight: 136.58
Purity/Analysis Method: >96.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 136
Flash Point (°C): 110
Freezing point (°C): 18
Specific Gravity (20/20): 1.17


Other Product Names / Synonyms: 5-Chloropentanoic Acid

Formula: C₅H₉ClO₂
Boiling Pt: 136 °C
Melting Pt: 17…19 °C
Density: 1.17 (20 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00004415
CAS Number: 1119-46-6
EINECS: 214-279-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=215bee66c731de173c2a6ccca6b38c240a0e94a9&vid=215bee66c731de173c2a6ccca6b38c240a0e94a9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC,T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR