meso-Butane-1,2,3,4-tetracarboxylic acid ≥98.0%
Supplier: TCI America
Synonyms:
trans-Butane-1,2,3,4-tetracarboxylic acid
B0684-25G
B0684-500G
TCB0684-025GEA
47.4
CAD
TCB0684-025G
TCB0684-500G
meso-Butane-1,2,3,4-tetracarboxylic acid ≥98.0%
meso-Butane-1,2,3,4-tetracarboxylic acid
CAS Number: 4534-68-3
MDL Number: MFCD00070496
Molecular Formula: C8H10O8
Molecular Weight: 234.16
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Formula:
C₈H₁₀O₈ |
MDL Number:
MFCD00070496 CAS Number: 4534-68-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...