2,3-Butanediol ≥97.0% for synthesis

Supplier: TCI America

Synonyms: 2,3-Dihydroxy butane, (±)-2,3-Butanediol, 2,3-Butylene glykol

B0681-500G B0681-25G B0681-100G
TCB0681-500GEA 540.36 CAD
TCB0681-500G TCB0681-025G 76219-324
2,3-Butanediol ≥97.0% for synthesis
2,3-Butanediol

CAS Number: 513-85-9
MDL Number: MFCD00004523
Molecular Formula: C4H10O2
Molecular Weight: 90.12
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 182
Flash Point (°C): 85
Specific Gravity (20/20): 0.99


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 2,3-Butylene Glycol, 2,3-Dihydroxybutane

Formula: C₄H₁₀O₂
Boiling Pt: 182 °C
Melting Pt: 18…20 °C
Density: 0.99 (20 °C)
Flash Pt: 85 °C
Storage Temperature: Ambient
MDL Number: MFCD00004523
CAS Number: 513-85-9
EINECS: 208-173-6
Merck Index: 12,01602
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9651d1b05bc5f743df84adb0f5418a08d2a1cd54&vid=5446b2be9f5547fd9fcf7346856312ac9c81aad1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR