6-Amino-1-naphthalenesulfonic acid ≥97.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
6-Amino-1-naphthalenesulphonic acid, 6-Amino-1-naphthalenesulfonic acid
A0347-25G
A0347-500G
TCA0347-025GEA
104.12
CAD
TCA0347-025G
TCA0347-500G
6-Amino-1-naphthalenesulfonic acid ≥97.0% (by HPLC, titration analysis)
6-Amino-1-naphthalenesulphonic acid
CAS Number: 81-05-0
MDL Number: MFCD00035724
Molecular Formula: C10H9NO3S
Molecular Weight: 223.25
Purity/Analysis Method: >97.0% (HPLC,T)
Form: Crystal
Other Product Names / Synonyms: Dahl's Acid, 2-Naphthylamine-5-sulfonic Acid
Formula:
C₁₀H₉NO₃S MW: 223.2500 g/mol |
MDL Number:
MFCD00035724 CAS Number: 81-05-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...