Tri-o-tolyl borate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Boric Acid Tri-o-cresyl Ester, Boric Acid Tri-o-tolyl Ester, Tri-o-cresyl Borate

B0519-5ML B0519-25ML
TCB0519-5MLEA 197.5 CAD
TCB0519-5ML TCB0519-025ML
Tri-o-tolyl borate ≥98.0% (by titrimetric analysis)
Tri-o-tolyl borate

CAS Number: 2665-12-5
MDL Number: MFCD00059311
Molecular Formula: C21H21BO3
Molecular Weight: 332.21
Purity/Analysis Method: >98.0% (T)
Form: Clear Liquid
Boiling point (°C): 386
Flash Point (°C): 174
Specific Gravity (20/20): 1.09


Other Product Names / Synonyms: Boric Acid Tri-o-cresyl Ester, Tri-o-cresyl Borate, Boric Acid Tri-o-tolyl Ester

Formula: C₂₁H₂₁BO₃
MW: 332.21 g/mol
Boiling Pt: 386 °C
Density: 1.09 (20 °C)
Flash Pt: 174 °C
MDL Number: MFCD00059311
CAS Number: 2665-12-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5a924849b2ca12fba9c1617d1cf23298530d60ef&vid=5a924849b2ca12fba9c1617d1cf23298530d60ef&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR