4,4'-Isopropylidenediphenol ≥99.0% for synthesis
Supplier: TCI America
Synonyms:
Bisphenol A, BPA, 2,2-Bis(4-hydroxyphenyl)propane
CAS Number: 80-05-7
MDL Number: MFCD00002366
Molecular Formula: C15H16O2
Molecular Weight: 228.29
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 252
Melting point (°C): 156
Flash Point (°C): 213
Product Categories: Materials Chemistry, Monomers, Bisphenol A type Compounds (for High-Performance Polymer Research), Pressure & Heat Sensitive Recording Materials, Analytical Science, Bisphenol A (Environmental Endocrine Disruptors)
Other Product Names / Synonyms: Bisphenol A, 4,4'-Isopropylidenediphenol
Formula:
C₁₅H₁₆O₂ MW: 228.29 g/mol Boiling Pt: 401 Melting Pt: 155 Density: 1.205 g/cm³ (20 °C) Flash Pt: 213 °C |
MDL Number:
MFCD00002366 CAS Number: 80-05-7 EINECS: 201-245-8 Merck Index: 13,01296 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...