3,6-Bis(hydroxymethyl)durene ≥99.0%

Supplier: TCI America

Synonyms: 2,3,5,6-Tetramethyl-p-xylene-alpha,alpha'-diol, 1,4-Bis(hydroxymethyl)-2,3,5,6-tetramethylbenzene

B0493-5G B0493-1G
TCB0493-5GEA 502.44 CAD
TCB0493-5G TCB0493-001G
3,6-Bis(hydroxymethyl)durene ≥99.0%
3,6-Bis(hydroxymethyl)durene

CAS Number: 7522-62-5
MDL Number: MFCD00004623
Molecular Formula: C12H18O2
Molecular Weight: 194.27
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Melting point (°C): 256


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 1,4-Bis(hydroxymethyl)-2,3,5,6-tetramethylbenzene, 2,3,5,6-Tetramethyl-p-xylene-alpha,alpha'-diol

Formula: C₁₂H₁₈O₂
MW: 194.27 g/mol
Melting Pt: 256 °C
MDL Number: MFCD00004623
CAS Number: 7522-62-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=93d29ef5d97df30989e772363cd16e521c65d858&vid=93d29ef5d97df30989e772363cd16e521c65d858&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR