Copper(II) bis(2-hydroxyethyl)dithiocarbamate ≥98.0%
Supplier: TCI America
Synonyms:
Bis(2-hydroxyethyl)dithiocarbamic Acid Copper(II) Salt
B0491-25G
B0491-5G
TCB0491-025GEA
247.9
CAD
TCB0491-025G
TCB0491-5G
Copper(II) bis(2-hydroxyethyl)dithiocarbamate ≥98.0%
Copper(II) bis(2-hydroxyethyl)dithiocarbamate
CAS Number: 52611-57-1
MDL Number: MFCD00059149
Molecular Formula: C5H11NO2S2
Molecular Weight: 424.07
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Product Categories: Materials Chemistry, Magnetic Metal Complexes (Magnetic Materials)
Other Product Names / Synonyms: Bis(2-hydroxyethyl)dithiocarbamic Acid Copper(II) Salt
Formula:
C₅H₁₁NO₂S₂ MW: 424.07 g/mol |
MDL Number:
MFCD00059149 CAS Number: 52611-57-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...