Bindschedler's green leuco base ≥98.0%
Supplier: TCI America
Synonyms:
Bis(4-dimethylaminophenyl)amine, Leuco Bindschedler's Green, N,N,N',N'-Tetramethyldiaminodiphenylamine
B0482-1G
B0482-5G
TCB0482-001GEA
388.68
CAD
TCB0482-001G
TCB0482-5G
Bindschedler's green leuco base ≥98.0%
Bindschedler's green leuco base
CAS Number: 637-31-0
MDL Number: MFCD00059291
Molecular Formula: C16H21N3
Molecular Weight: 255.37
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 121
Other Product Names / Synonyms: Bis(4-dimethylaminophenyl)amine, Leuco Bindschedler's Green, N,N,N',N'-Tetramethyldiaminodiphenylamine
Formula:
C₁₆H₂₁N₃ Melting Pt: 121 °C |
MDL Number:
MFCD00059291 CAS Number: 637-31-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...