D-(+)-Biotin ≥98.0% for synthesis

Supplier: TCI America

Synonyms: Vitamin H, D-Biotin, Vitamin B8, 5-((3aS,4S,6aR)-2-Oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanoic acid, Vitamin B7

B0463-5G B0463-1G B0463-100MG
TCB0463-5GEA 467.82 CAD
TCB0463-5G TCB0463-001G TCB0463-100MG
D-(+)-Biotin ≥98.0% for synthesis
D(+)-Biotin

CAS Number: 58-85-5
MDL Number: MFCD00005541
Molecular Formula: C10H16N2O3S
Molecular Weight: 244.31
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 230
Specific rotation [a]20/D: 90.5 deg (C=2, 0.1mol/L NaOH sol.)


Product Categories: Bioscience, Vitamins, Biotins & Streptavidins
Other Product Names / Synonyms: Vitamin H

Formula: C₁₀H₁₆N₂O₃S
Melting Pt: 230 °C
Density: 1.41 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00005541
CAS Number: 58-85-5
EINECS: 200-399-3
Merck Index: 13,01231
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=be403c0cca0d612de378d076e6027bf290c90493&vid=be403c0cca0d612de378d076e6027bf290c90493&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR