Girard's reagent T (carbazoylmethyltrimethylammonium chloride) ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 2-Hydrazinyl-N,N,N-trimethyl-2-oxoethanaminium chloride

B0457-25G B0457-500G
TCB0457-025GEA 44.98 CAD
TCB0457-025G TCB0457-500G
Girard's reagent T (carbazoylmethyltrimethylammonium chloride) ≥98.0% (by titrimetric analysis)
Girard's reagent T (Carbazoylmethyltrimethylammonium chloride)

CAS Number: 123-46-6
MDL Number: MFCD00012009
Molecular Formula: C5H14ClN3O
Molecular Weight: 167.64
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 190


Product Categories: Analytical Science, Derivatization Reagents for MS
Other Product Names / Synonyms: Betaine Hydrazide Hydrochloride, (Hydrazinocarbonylmethyl)trimethylammonium Chloride, Trimethylacetohydrazideammonium Chloride

Formula: C₅H₁₄ClN₃O
MW: 167.64 g/mol
Melting Pt: 190 °C
MDL Number: MFCD00012009
CAS Number: 123-46-6
EINECS: 204-629-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bb6a70d7bc4ba8944f6e07365e918fb4307b57e8&vid=bb6a70d7bc4ba8944f6e07365e918fb4307b57e8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR