Beryllon III

Supplier: TCI America

Synonyms: 5-(4-Diethylamino-2-hydroxyphenylazo)-4-hydroxynaphthalene-2,7-disulphonic Acid Sodium Salt, 5-(4-Diethylamino-2-hydroxyphenylazo)-4-hydroxynaphthalene-2,7-disulfonic Acid Sodium Salt

B0453-5G B0453-1G
TCB0453-5GEA 317.42 CAD
TCB0453-5G TCB0453-001G
Beryllon III
Beryllon III

CAS Number: 3627-04-1
MDL Number: MFCD00070629
Molecular Formula: C20H21N3O8S2
Molecular Weight: 517.50
Form: Crystal
Color: Deep Green


Other Product Names / Synonyms: 5-(4-Diethylamino-2-hydroxyphenylazo)-4-hydroxynaphthalene-2,7-disulfonic Acid Sodium Salt

Formula: C₂₀H₂₁N₃O₈S₂
MW: 517.50 g/mol
MDL Number: MFCD00070629
CAS Number: 3627-04-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=34b6fcd93927e277ba84cdc54cbe106b9d308182&vid=34b6fcd93927e277ba84cdc54cbe106b9d308182&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR