2-(Benzyloxy)ethanol ≥98.0%

Supplier: TCI America

B0426-25ML B0426-250ML
TCB0426-025MLEA 67.78 CAD
TCB0426-025ML TCB0426-250ML
2-(Benzyloxy)ethanol ≥98.0%
2-(Benzyloxy)ethanol

CAS Number: 622-08-2
MDL Number: MFCD00002868
Molecular Formula: C9H12O2
Molecular Weight: 152.19
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 256
Flash Point (°C): 130
Specific Gravity (20/20): 1.07


Other Product Names / Synonyms: Benzyl Cellosolve, Benzyl Glycol, Ethylene Glycol Monobenzyl Ether

Formula: C₉H₁₂O₂
Boiling Pt: 256 °C
Density: 1.07 (20 °C)
Flash Pt: 130 °C
MDL Number: MFCD00002868
CAS Number: 622-08-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=26467f4aeb1d68f57d63ebdc026a7bf98b695cb6&vid=26467f4aeb1d68f57d63ebdc026a7bf98b695cb6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR