Benzyl orange
Supplier: TCI America
B0425-5G
B0425-1G
TCB0425-005GEA
378.57
CAD
TCB0425-005G
TCB0425-001G
Benzyl orange
Benzyl orange
CAS Number: 36402-77-4
MDL Number: MFCD00198097
Molecular Formula: C19H17N3O3S
Molecular Weight: 389.40
Form: Crystal
Color: Reddish Yellow
Formula:
C₁₉H₁₇N₃O₃S |
MDL Number:
MFCD00198097 CAS Number: 36402-77-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...