Benzoyl-DL-valine ≥98.0% (by titrimetric analysis)

Supplier: TCI America

B0401-25G B0401-1G
TCB0401-25GEA 439.71 CAD
TCB0401-25G TCB0401-001G
Benzoyl-DL-valine ≥98.0% (by titrimetric analysis)
Benzoyl-DL-valine

CAS Number: 2901-80-6
MDL Number: MFCD00038283
Molecular Formula: C12H15NO3
Molecular Weight: 221.26
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 132

Formula: C₁₂H₁₅NO₃
Melting Pt: 132 °C
MDL Number: MFCD00038283
CAS Number: 2901-80-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5269d12aa993f01933016f2b9bde18c77c13134d&vid=5269d12aa993f01933016f2b9bde18c77c13134d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR