Silver behenate ≥95.0% (by titrimetric analysis)

Supplier: TCI America

B0398-5G B0398-25G
TCB0398-5GEA 167.48 CAD
TCB0398-5G TCB0398-025G
Silver behenate ≥95.0% (by titrimetric analysis)
Silver behenate

CAS Number: 2489-05-6
MDL Number: MFCD00059001
Molecular Formula: C22H44O2
Molecular Weight: 447.45
Purity/Analysis Method: >95.0% (T)
Form: Crystal


Other Product Names / Synonyms: Behenic Acid Silver Salt, Docosanoic Acid Silver Salt, Silver Docosanoate

Formula: C₂₂H₄₄O₂
MDL Number: MFCD00059001
CAS Number: 2489-05-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3cefb77632256beb0bcd2c3106afcde5ad1a5ee9&vid=3cefb77632256beb0bcd2c3106afcde5ad1a5ee9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR