2,2'-Bibenzothiazoline ≥85.0% (by titrimetric analysis)

Supplier: TCI America

B0377-5G B0377-1G
TCB0377-5GEA 181.86 CAD
TCB0377-5G TCB0377-001G
2,2'-Bibenzothiazoline ≥85.0% (by titrimetric analysis)
2,2'-Bibenzothiazoline

CAS Number: 19258-20-9
MDL Number: MFCD00059699
Molecular Formula: C14H12N2S2
Molecular Weight: 272.38
Purity/Analysis Method: >85.0% (T)
Form: Crystal
Melting point (°C): 135


Other Product Names / Synonyms: 2,2'-Bisbenzothiazoline, 2,2'-Dibenzothiazoline

Formula: C₁₄H₁₂N₂S₂
Melting Pt: 135 °C
MDL Number: MFCD00059699
CAS Number: 19258-20-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=397ec05bd7f48c8b4ea6819b88eec6306daf029a&vid=397ec05bd7f48c8b4ea6819b88eec6306daf029a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 85.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR