Monopotassium 7-Amino-1,3-naphthalenedisulfonate Hydrate ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: Potassium Hydrogen 7-Amino-1,3-naphthalenedisulphonate, 2-Naphthylamine-6,8-disulfonic acid Monopotassium salt, 7-Amino-1,3-naphthalenedisulfonic acid Monopotassium salt, 7-Amino-1,3-naphthalenedisulphonic acid Monopotassium salt, Potassium Hydrogen 7-Amino-1,3-naphthalenedisulfonate, 2-Naphthylamine-6,8-disulphonic acid Monopotassium salt, Amino G acid Monopotassium salt

A0337-25G A0337-500G
TCA0337-025GEA 61.07 CAD
TCA0337-025G TCA0337-500G
Monopotassium 7-Amino-1,3-naphthalenedisulfonate Hydrate ≥98.0% (by HPLC)
Monopotassium 7-Amino-1,3-naphthalenedisulfonate Hydrate

CAS Number: 842-15-9
MDL Number: MFCD00064177
Molecular Formula: C10H9NO6S2
Molecular Weight: 341.39
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal


Other Product Names / Synonyms: Amino G Acid Monopotassium Salt, 7-Amino-1,3-naphthalenedisulfonic Acid Monopotassium Salt, 2-Naphthylamine-6,8-disulfonic Acid Monopotassium Salt, Potassium Hydrogen 7-Amino-1,3-naphthalenedisulfonate

Formula: C₁₀H₉NO₆S₂
MW: 341.39 g/mol
MDL Number: MFCD00064177
CAS Number: 842-15-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9cdca56d94c94db8fe66c4d6653773730ea6ef57&vid=9cdca56d94c94db8fe66c4d6653773730ea6ef57&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR