2-Amino-4-methylthiazole hydrochloride ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

A0336-5G A0336-25G
TCA0336-005GEA 45.82 CAD
TCA0336-005G TCA0336-025G
2-Amino-4-methylthiazole hydrochloride ≥98.0% (by HPLC, total nitrogen)
2-Amino-4-methylthiazole hydrochloride

CAS Number: 6142-15-0
MDL Number: MFCD00035214
Molecular Formula: C4H6N2S
Molecular Weight: 150.62
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 173

Formula: C₄H₆N₂S
Melting Pt: 173 °C
MDL Number: MFCD00035214
CAS Number: 6142-15-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=766ec423c484599ae6932d41e6eecfe28a62d702&vid=766ec423c484599ae6932d41e6eecfe28a62d702&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR