2,3-Diphenyl-5-(2-thienyl)tetrazolium chloride ≥97.0%

Supplier: TCI America

Synonyms: STC

B0325-1G B0325-100MG
TCB0325-001GEA 647.96 CAD
TCB0325-001G TCB0325-100MG
2,3-Diphenyl-5-(2-thienyl)tetrazolium chloride ≥97.0%
2,3-Diphenyl-5-(2-thienyl)tetrazolium chloride

CAS Number: 38800-20-3
MDL Number: MFCD00060088
Molecular Formula: C17H13ClN4S
Molecular Weight: 340.83
Purity/Analysis Method: >97.0% (HPLC,T)
Form: Crystal


Other Product Names / Synonyms: STC

Formula: C₁₇H₁₃ClN₄S
MW: 340.83 g/mol
MDL Number: MFCD00060088
CAS Number: 38800-20-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=75f3c2fd53f2b829d86232b8c3b96c3720b69aa7&vid=75f3c2fd53f2b829d86232b8c3b96c3720b69aa7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR