N-Benzoyl-DL-phenylalanine-2-naphthyl ester ≥98.0% (by HPLC)

Supplier: TCI America

B0309-1G B0309-100MG
TCB0309-001GEA 330.22 CAD
TCB0309-001G TCB0309-100MG
N-Benzoyl-DL-phenylalanine-2-naphthyl ester ≥98.0% (by HPLC)
N-Benzoyl-DL-phenylalanine-2-naphthyl ester

[for Determination of Chymotrypsin]
CAS Number: 15873-25-3
MDL Number: MFCD00021607
Molecular Formula: C26H21NO3
Molecular Weight: 395.46
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 158

Formula: C₂₆H₂₁NO₃
Melting Pt: 158 °C
MDL Number: MFCD00021607
CAS Number: 15873-25-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e5e0bb1d98e9bd1bb98134ab2a57fb5bb62215a1&vid=e5e0bb1d98e9bd1bb98134ab2a57fb5bb62215a1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR