Benzoylthiocholine iodide ≥98.0%

Supplier: TCI America

B0307-1G B0307-5G B0307-25G
TCB0307-001GEA 99.54 CAD
TCB0307-001G TCB0307-005G TCB0307-25G
Benzoylthiocholine iodide ≥98.0%
Benzoylthiocholine iodide

CAS Number: 10561-14-5
MDL Number: MFCD00059970
Molecular Formula: C12H18INOS
Molecular Weight: 351.25
Purity/Analysis Method: >98.0% (T)
Form: Crystal

Formula: C₁₂H₁₈INOS
MDL Number: MFCD00059970
CAS Number: 10561-14-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=93c299b53a203fe92eb75d1440298766d812414c&vid=93c299b53a203fe92eb75d1440298766d812414c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR