Glycine benzyl ester-p-toluenesulfonate ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: Benzyl glycinate 4-methylbenzenesulfonate, Benzyl glycinate 4-methylbenzenesulphonate, H-Gly-OBzl.Tos-OH, H-Gly-OBzl.TosOH

B0266-25G B0266-5G
TCB0266-025GEA 58.46 CAD
TCB0266-025G TCB0266-5G
Glycine benzyl ester-p-toluenesulfonate ≥98.0% (by HPLC, titration analysis)
Glycine benzyl ester-p-toluenesulfonate

CAS Number: 1738-76-7
MDL Number: MFCD00035425
Molecular Formula: C9H11NO2
Molecular Weight: 337.39
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 135


Product Categories: Bioscience, Amino Acids (C-Protected)
Other Product Names / Synonyms: H-Gly-OBzl.Tos-OH

Formula: C₉H₁₁NO₂
MW: 337.39 g/mol
Melting Pt: 135 °C
MDL Number: MFCD00035425
CAS Number: 1738-76-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bb795b8aa5c0b75df66e9fa7334d3de59eec6b93&vid=bb795b8aa5c0b75df66e9fa7334d3de59eec6b93&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR