Benzoyl leuco methylene blue ≥96.0% (by HPLC, titration analysis)

Supplier: TCI America

B0207-1G B0207-25G
TCB0207-001GEA 53.72 CAD
TCB0207-001G TCB0207-025G
Benzoyl leuco methylene blue ≥96.0% (by HPLC, titration analysis)
Benzoyl leuco methylene blue

CAS Number: 1249-97-4
MDL Number: MFCD00059153
Molecular Formula: C23H23N3OS
Molecular Weight: 389.52
Purity/Analysis Method: >96.0% (HPLC,T)
Form: Crystal
Melting point (°C): 198

Formula: C₂₃H₂₃N₃OS
Melting Pt: 198 °C
MDL Number: MFCD00059153
CAS Number: 1249-97-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e8a8ff2bdebcd9b8730fb3cf353c8f8f197d54ec&vid=e8a8ff2bdebcd9b8730fb3cf353c8f8f197d54ec&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (HPLC,T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR