Benzoyl-DL-leucine ≥99.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: Bz-DL-Leu-OH

B0206-10G B0206-25G
TCB0206-010GEA 48.98 CAD
TCB0206-010G TCB0206-025G
Benzoyl-DL-leucine ≥99.0% (by HPLC, titration analysis)
Benzoyl-DL-leucine

CAS Number: 17966-67-5
MDL Number: MFCD00038295
Molecular Formula: C13H17NO3
Molecular Weight: 235.28
Purity/Analysis Method: >99.0% (HPLC,T)
Form: Crystal
Melting point (°C): 141

Formula: C₁₃H₁₇NO₃
MW: 235.28 g/mol
Melting Pt: 141 °C
MDL Number: MFCD00038295
CAS Number: 17966-67-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6b1e99ab997e0624e1f16e44964a90b1dccc127f&vid=6b1e99ab997e0624e1f16e44964a90b1dccc127f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR