Benzoyl-L-histidine monohydrate ≥98.0%
Supplier: TCI America
Synonyms:
Bz-His-OH
B0204-1G
B0204-100MG
TCB0204-001GEA
64.78
CAD
TCB0204-001G
TCB0204-100MG
Benzoyl-L-histidine monohydrate ≥98.0%
Benzoyl-L-histidine monohydrate
CAS Number: 5354-94-9
MDL Number: MFCD00037850
Molecular Formula: C13H13N3O3
Molecular Weight: 259.27
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Specific rotation [a]20/D: -47 deg (C=2, 0.5mol/L HCl)
Formula:
C₁₃H₁₃N₃O₃ MW: 259.27 g/mol |
MDL Number:
MFCD00037850 CAS Number: 5354-94-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...