N-Benzoyl-L-glutamic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

B0202-1G B0202-25G
TCB0202-001GEA 28.12 CAD
TCB0202-001G TCB0202-025G
N-Benzoyl-L-glutamic acid ≥98.0% (by HPLC, titration analysis)
N-Benzoyl-L-glutamic acid

CAS Number: 6094-36-6
MDL Number: MFCD00014028
Molecular Formula: C12H13NO5
Molecular Weight: 251.24
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 140
Specific rotation [a]20/D: 18.5 deg (C=10, 1mol/L NaOH sol.)

Formula: C₁₂H₁₃NO₅
Melting Pt: 140 °C
MDL Number: MFCD00014028
CAS Number: 6094-36-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=52a9eb271f12819bdfc9484ecee35164c460ad0f&vid=52a9eb271f12819bdfc9484ecee35164c460ad0f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR