N-Benzoyl-D-glucosamine ≥98.0%

Supplier: TCI America

Synonyms: N-((2R,3R,4S,5R)-3,4,5,6-Tetrahydroxy-1-oxohexan-2-yl)benzamide, N-Benzoyl-D-glucosamine, 2-Benzamido-2-deoxy-D-glucopyranose

B0200-25G B0200-5G
TCB0200-025GEA 153.26 CAD
TCB0200-025G TCB0200-5G
N-Benzoyl-D-glucosamine ≥98.0%
N-Benzoyl-D-glucosamine

CAS Number: 655-42-5
MDL Number: MFCD00070521
Molecular Formula: C13H17NO6
Molecular Weight: 283.28
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Specific rotation [a]20/D: 43 deg (C=2, H2O)


Product Categories: Bioscience, Glucosamine, Aminosugars
Other Product Names / Synonyms: 2-Benzamido-2-deoxy-D-glucopyranose

Formula: C₁₃H₁₇NO₆
MW: 283.28 g/mol
MDL Number: MFCD00070521
CAS Number: 655-42-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=28c0e29f227bb039101f56d5049a14837e938ee7&vid=28c0e29f227bb039101f56d5049a14837e938ee7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR