Saccharin sodium dihydrate ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
B0131-25G
B0131-500G
TCB0131-025GEA
58.46
CAD
TCB0131-025G
TCB0131-500G
Saccharin sodium dihydrate ≥98.0% (by HPLC, titration analysis)
Saccharin sodium dihydrate
CAS Number: 6155-57-3
MDL Number: MFCD00151213
Molecular Formula: C7H5NO3S
Molecular Weight: 205.16
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 228
Product Categories: Bioscience, Obesity/Diabete Research Reagents
Other Product Names / Synonyms: o-Benzoic Sulfimide Sodium Salt, Saccharin Sodium Salt
Formula:
C₇H₅NNaO₃S·2H₂O Melting Pt: 228 °C |
MDL Number:
MFCD00151213 CAS Number: 6155-57-3 EINECS: 204-886-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...