2-Ethylhexyl butyrate ≥98.0%
Supplier: TCI America
B0130-500ML
B0130-25ML
TCB0130-500MLEA
532.46
CAD
TCB0130-500ML
TCB0130-025ML
2-Ethylhexyl butyrate ≥98.0%
2-Ethylhexyl butyrate
CAS Number: 25415-84-3
MDL Number: MFCD00059428
Molecular Formula: C12H24O2
Molecular Weight: 200.32
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 237
Specific Gravity (20/20): 0.87
Other Product Names / Synonyms: Butyric Acid Octyl Ester, Butyric Acid 2-Ethylhexyl Ester, Octyl Butyrate
Formula:
C₁₂H₂₄O₂ Boiling Pt: 237 °C Density: 0.87 (20 °C) |
MDL Number:
MFCD00059428 CAS Number: 25415-84-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...