α-N-Benzoyl-L-argininamide hydrochloride monohydrate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Bz-Arg-NH2 HClBAAN-α-Benzoyl-L-argininamide hydrochloride monohydrateNardis Reagent

B0102-1G B0102-25G
TCB0102-025GEA 587.49 CAD
TCB0102-025G TCB0102-001G
α-N-Benzoyl-L-argininamide hydrochloride monohydrate ≥98.0% (by titrimetric analysis)
α-N-Benzoyl-L-argininamide hydrochloride monohydrate

Form: Crystal
Specific rotation [a]20/D: 6.5 deg (C=4, H2O)

Formula: C₁₃H₁₉N₅O₂·HCl·H₂O
MW: 313.79 g/mol (anhy)
MDL Number: MFCD00035003
CAS Number: 4299-03-0
https://ca.vwr.com/stibo/web/std.lang.all/22/61/28932261.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR