5-(Benzoylamino)valeric acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

B0101-100MG B0101-5G B0101-1G
TCB0101-100MGEA 48.98 CAD
TCB0101-100MG TCB0101-5G TCB0101-001G
5-(Benzoylamino)valeric acid ≥98.0% (by titrimetric analysis)
5-(Benzoylamino)valeric acid

CAS Number: 15647-47-9
MDL Number: MFCD00021790
Molecular Formula: C12H15NO3
Molecular Weight: 221.26
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 94

Formula: C₁₂H₁₅NO₃
MW: 221.26 g/mol
Melting Pt: 94 °C
MDL Number: MFCD00021790
CAS Number: 15647-47-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0537e42364971e566781028aba254ef9217a4556&vid=0537e42364971e566781028aba254ef9217a4556&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR