α-Benzoinoxime ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

B0081-25G B0081-500G
TCB0081-025GEA 97.96 CAD
TCB0081-025G TCB0081-500G
α-Benzoinoxime ≥98.0% (by HPLC, total nitrogen)
α-Benzoinoxime

CAS Number: 441-38-3
MDL Number: MFCD00004501
Molecular Formula: C14H13NO2
Molecular Weight: 227.26
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 153


Other Product Names / Synonyms: Cupron

Formula: C₁₄H₁₃NO₂
Melting Pt: 153 °C
MDL Number: MFCD00004501
CAS Number: 441-38-3
EINECS: 207-127-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=69306df090b6b9a3d73b63ff302abe8b0178be52&vid=69306df090b6b9a3d73b63ff302abe8b0178be52&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR