Isopentyl benzoate ≥98.0%

Supplier: TCI America

B0071-500G B0071-25G
TCB0071-500GEA 173.8 CAD
TCB0071-500G TCB0071-025G
Isopentyl benzoate ≥98.0%
Isopentyl benzoate

CAS Number: 94-46-2
MDL Number: MFCD00026515
Molecular Formula: C12H16O2
Molecular Weight: 192.26
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 262
Flash Point (°C): 129
Specific Gravity (20/20): 0.99


Other Product Names / Synonyms: Benzoic Acid Isoamyl Ester, Isopentyl Benzoate

Formula: C₁₂H₁₆O₂
Boiling Pt: 262 °C
Density: 0.99 (20 °C)
Flash Pt: 129 °C
Storage Temperature: Ambient
MDL Number: MFCD00026515
CAS Number: 94-46-2
EINECS: 202-334-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f13cc0c02c3f0ed6221cbfc50ebac6f5fe675e41&vid=b98ef0064a2f8d11023f483159c4ce7a5c44b317&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR