Ethyl benzenesulfonate ≥98.0%

Supplier: TCI America

Synonyms: Benzenesulfonic Acid Ethyl Ester, Benzenesulphonic Acid Ethyl Ester

B0032-500G B0032-25G B0032-100G
TCB0032-500GEA 717.32 CAD
TCB0032-500G TCB0032-025G TCB0032-100G
Ethyl benzenesulfonate ≥98.0%
Ethyl benzenesulfonate

CAS Number: 515-46-8
MDL Number: MFCD00025033
Molecular Formula: C8H10O3S
Molecular Weight: 186.23
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 156
Specific Gravity (20/20): 1.22


Other Product Names / Synonyms: Benzenesulfonic Acid Ethyl Ester

Formula: C₈H₁₀O₃S
MW: 186.23 g/mol
Boiling Pt: 156 °C
Density: 1.22 (20 °C)
MDL Number: MFCD00025033
CAS Number: 515-46-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f6f4e20b83edda12233a24414e596f9deb424fbd&vid=f6f4e20b83edda12233a24414e596f9deb424fbd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR