Benzenesulfonanilide ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

B0029-25G B0029-500G
TCB0029-025GEA 127.98 CAD
TCB0029-025G TCB0029-500G
Benzenesulfonanilide ≥98.0% (by HPLC, total nitrogen)
Benzenesulfonanilide

CAS Number: 1678-25-7
MDL Number: MFCD00035919
Molecular Formula: C12H11NO2S
Molecular Weight: 233.29
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 110

Formula: C₁₂H₁₁NO₂S
Melting Pt: 110 °C
MDL Number: MFCD00035919
CAS Number: 1678-25-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=437d452392c176e42a0aa33229fe4099f66a17ad&vid=437d452392c176e42a0aa33229fe4099f66a17ad&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR