Benzenesulfonamide ≥98.0% (by total nitrogen basis)

Supplier: TCI America
Warning

B0028-500G B0028-25G
TCB0028-500GEA 357.08 CAD
TCB0028-500G TCB0028-025G
Benzenesulfonamide ≥98.0% (by total nitrogen basis)
Benzenesulphonamide

CAS Number: 98-10-2
MDL Number: MFCD00007930
Molecular Formula: C6H7NO2S
Molecular Weight: 157.19
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Melting point (°C): 154
Flash Point (°C): 250

Formula: C₆H₇NO₂S
MW: 157.19 g/mol
Melting Pt: 154 °C
Flash Pt: 250 °C
MDL Number: MFCD00007930
CAS Number: 98-10-2
EINECS: 202-637-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7ccd4216e30fd98811217ce8a54b1a2a0d1842c7&vid=7ccd4216e30fd98811217ce8a54b1a2a0d1842c7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR